Nichipet ECO Glass Tip Pipettors, Nichiryo America
Supplier: Nichiryo America
00-NPC-1000 00-NPC-5000 00-NPC-200 00-NPC-10ML
89078-458EA 772.62 CAD
89078-458 89078-460 89078-456 89078-462
Nichipet ECO Glass Tip Pipettors, Nichiryo America
Pipettes Micropipettes
Designed to provide the accuracy and precision of a micropipettor with the compatibility of glass, the unique Nichipet ECO utilizes reliable and stable glass tips to dispense liquids quickly and safely.
  • Pipettors feature molded channels within the handle for improved grip
  • Glass tips are autoclavable
  • Four different pipettor sizes cover a volume range from 20 µl to 1 ml
Tips are attached to the unit with a double O-ring design, ensuring a secure fit with highly reproducible results. Volume is adjustable through the use of a color-coded dial which locksMore Product Information