Nichitip Pipette Tips for Nichiryo Pipettes
Supplier: Nichiryo America
BMT-X BMT-Z BMT-XR BMT-XRS BMT-ZR BMT-ZRS
89078-452PK 73.8 CAD
89078-452 89078-454 76808-738 76808-758 76808-740 76808-742
Nichitip Pipette Tips for Nichiryo Pipettes
Pipette Tips
These tips are made of 100% virgin polypropyleylene.
  • High quality
  • Free of detectable DNase, RNase, DNA
  • Autoclavable
Made in the highest quality moulds, these tips can be used whenever the work demands a higher standard.