Ovation® ESC, Electronic Single Channel Pipettors, VistaLab
Supplier: VistaLab
1065-0020 1065-0250 1065-1250 1065-0125
89005-058EA 1971.62 CAD
89005-058 89005-062 89005-064 89005-060
Ovation® ESC, Electronic Single Channel Pipettors, VistaLab
Pipettes Micropipettes
These adjustable volume, air displacement, fully electronic motor-driven pipettors aspirate and dispense precise fluid volumes.
  • Acquire tips with only a gentle push; audible click ensures a secure seal
  • Contoured plunger electronically activates pipetting activity
Pipettors feature LCD display and single keystroke pushbutton controls for easy access to liquid handling functions, calibration, settings, status, and remaining battery power. Energy transfer button uses energy captured during tip acquisition to eject tip with minimal thumb force.