Panel-Mounted Gooseneck Mixing Faucets, with Four-Arm Handles, WaterSaver Faucet
Supplier: WaterSaver Faucet
L514VB-WSA L514WSA L511WS L511VB-WS
VFL512VB-WSEA 946.83 CAD
VFL512VB-WS VFL512WS VFT-0354-06 VFT-0354-6V
Panel-Mounted Gooseneck Mixing Faucets, with Four-Arm Handles, WaterSaver Faucet
Faucets and Sinks Faucets
Panel-mounted mixing faucets are made of chrome-plated cast brass and provided with a serrated hose end, four-arm handles, and 1 cm (3/8") IPS inlet shanks, locknuts, and washers
Faucets are available with 15.2 cm (6") spread gooseneck, with or without a vacuum breaker. Faucet valves are self-contained renewable units with replaceable stainless steel seats.
Certifications: CSA certified. Comply with ANSI/ASME A112.18.1M.