DynaBloc Heating Blocks, Cylindrical, Ace Glass
Supplier: Ace Glass
13698-14 13698-49 13698-05 13698-20 13698-30 13698-21 13698-12 13698-13 13698-41 13698-37 13698-38 13699-06 13699-03 13699-04 13699-09 13697-04 13697-08 13697-16 13697-40 13696-03 13696-07 13696-11 13698-03
89083-366EA 520.43 CAD
89083-366 89083-368 89083-370 89083-372 89083-374 89083-376 89083-378 89083-380 89083-382 89083-384 89083-386 89083-388 89083-390 89083-392 89083-394 89083-396 89083-398 89083-400 89083-402 89083-404 89083-406 89083-408 89097-314
DynaBloc Heating Blocks, Cylindrical, Ace Glass
Block Heaters Blocks
These anodized aluminum blocks are ideal for heating and mixing solutions in a variety of vials, tubes, and flasks for applications such as digestion, extraction, distillation, and synthesis
Universal base plate is required for operating all DynaBloc cylindrical heating blocks and fits a standard, circular, 135mm diameter stirrer/hotplate top. Blocks can be separated from the base for easy cleaning and handling. The base has a hole in the side to accommodate a standard thermocouple sensor for heat measurement. Base plate dimensions: 135I.D.x164O.D.mm (55/16x67/16"). Blocks for vials andMore Product Information