Nalgene® Safety Waste Systems, Thermo Scientific
Supplier: Thermo Fisher Scientific
6379-0004 6379-0010
73520-202CS 395.02 CAD
73520-202 73520-204
Nalgene® Safety Waste Systems, Thermo Scientific
Bottles Solution Bottles
For temporary disposal of most chemical and biological wastes. Each system consists of a waste container, removable HDPE funnel and cover, polypropylene closure/funnel adapter, and PTFE vent membrane.
Red, hinged cover seals funnel, reducing the level of volatiles evaporating into the air. Easy-snap safety latch minimizes spillage if system is tipped over. Redesigned vent system with filter reduces fumes and splashing. Large-diameter funnels, in addition to the vents, enable fast and easy pouring. Funnel stem extends below the liquid level to reduce the evaporation rate. Removable neckMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable