Steam Valves, WaterSaver Faucet
Supplier: WaterSaver Faucet
L2892-146WS L2892-132SWS L2890-131WS L2892-134WS L2892-158WS L2892-132AWS L2892-131WS
59382-026EA 361.98 CAD
59382-026 59382-028 VFT-2890-00 59382-030 59382-032 59382-036 VFT-0304-01
Steam Valves, WaterSaver Faucet
Valves Air Regulation Valves
Straight pattern laboratory steam valve with removable angle serrated hose end.
  • Forged brass valve body
  • Fluoropolymer resin valve disc
  • Replaceable stainless steel seat
  • Black nylon heat-resistant four-arm handle
Maximum working pressure is 20 psi. Valve assemblies are furnished with 9.5 mm (3/8") IPS mounting shank, locknut and washer.