Socorex™ Calibra® 822 Digital Single-Channel Micropipettors, Variable Volume, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
851162 851163
80078-612CS 928.34 CAD
80078-612 80078-614 80078-616 80078-620 80078-622
Socorex™ Calibra® 822 Digital Single-Channel Micropipettors, Variable Volume, DWK Life Sciences
Pipettes Micropipettes
Sleek micropipettors are ideal for gel sequencing and biotechnology applications.
  • Micropipettors are also color-coded to ensure proper tip selection
  • Small sample volumes can be reproducibly dispensed
  • Autoclavable
They are equipped with twin cams with precalibrated steps to provide true digital volume entry, high performance, and calibration stability. Plunger assembly features fluoropolymer resin and stainless steel construction for a high degree of chemical resistance.
Ordering information: Supplied with a manufacturer’s three-year warranty onMore Product Information