Gas, Air, or Vacuum Fittings, Wall Mounted, WaterSaver Faucet
Supplier: WaterSaver Faucet
L4200FH158WS L4200-141WSA
97004-554EA 133.3 CAD
97004-554 97004-556
Gas, Air, or Vacuum Fittings, Wall Mounted, WaterSaver Faucet
Faucets and Sinks Faucets
These wall-mounted faucets are made of chrome-plated brass.
  • Maximum Working Pressure: 75 psi
  • Index Buttons for Gas, Air, and Vacuum
  • Single, Double, or Quadruple Service Available
  • Panel Flange or Wye Fitting
  • Chrome-Plated Brass Construction
They feature a panel flange or wye fitting and a ball valve for curb or wall mounting.
The faucet includes index buttons for gas, air, and vacuum, and single, double, or quadruple service is available.
Ordering information: This item includes 9.5 mm, (3/8") IPS tank nipples, locknuts, and washers.