Protein Molecular Weight Marker Mix, Rockland Immunchemicals
Supplier: Rockland Immunochemical
MB-201-0200 MB-201-1000 MB-201-2000
CAMB-201-0200EA 147 CAD
CAMB-201-0200 CAMB-201-1000 CAMB-201-2000
Protein Molecular Weight Marker Mix, Rockland Immunchemicals
Electrophoresis Reagents Molecular Weight Markers
This mixture contains seven purified proteins that range in size from 14 to 120kDa
The buffer composition is 62.5mM TRIS-HCl at pH 6.8, 2% (w/v) SDS, 250mM sodium chloride, 1mM EDTA, 4% (v/v) [beta]-mercaptoethanol, 35% (v/v) glycerol, and 0.05% (w/v) bromophenol blue with 0.01% (w/v) sodium azide as a preservative. Store marker mixture at –20°C (–4°F).
It has been formulated to yield well-defined bands after SDS-PAGE electrophoresis and staining with Coomassie blue or other common staining techniques.The bovine albumin band (67kDa) has been increased to serve as a reference. It is supplied in 1X loading buffer, so that it is ready to use.