Nalgene® Polycarbonate Straight-Sided Wide-Mouth Jars, Thermo Scientific
Supplier: Thermo Fisher Scientific
2116-0015 2116-0030 2116-0060 2116-0125 2116-0250 2116-0500 2116-1000
16129-300CS 329.9 CAD
16129-300 16129-302 16129-304 16129-306 16129-308 16129-310 16129-312
Nalgene® Polycarbonate Straight-Sided Wide-Mouth Jars, Thermo Scientific
Jars General Use Jars
Excellent for cold room and refrigerated use
Window-clear, polycarbonate with high impact resistance. Nontoxic, autoclavable, with a polypropylene screw closure. Not leakproof. Remove closure before autoclaving, or set on top of container at an angle without engaging threads.