Panel-Mounted Gooseneck Mixing Faucets, with Nylon Hooded Handles, WaterSaver Faucet
Supplier: WaterSaver Faucet
CT1714VB-WSA CT1714WS
VFCT1714VB-WSAEA 587.57 CAD
VFCT1714VB-WSA VFT-1714-00
Panel-Mounted Gooseneck Mixing Faucets, with Nylon Hooded Handles, WaterSaver Faucet
Faucets and Sinks Faucets
Panel-mounted mixing faucets are made of white epoxy/polyester hybrid powder-coated cast brass
They are provided with a serrated hose end, black nylon hooded handles, and 1 cm (3/8") mounting shanks, locknuts, and washers. Horizontal inlets are on 15.2 cm (6") centers and gooseneck spread is 15.2 cm (6"). Available with or without a vacuum breaker. Faucet valves are self-contained renewable units with replaceable stainless steel seats.
Certifications: CSA certified. Comply with ANSI/ASME A112.18.1M.