DNA (Deoxyribonucleic acid) solution (from calf thymus)
Supplier: Rockland Immunochemical
Convenient, ready-to-use liquid calf thymus DNA solution is designed for use in the preparation of pre-hybridization and hybridization solutions, and as a DNA carrier in yeast transformation protocols and other related methods
- Buffer composition: 10 mM Tris Chloride, 1 mM EDTA, ph 7.6
- Hyperchromocity: 31.4%
- Nitrogen: 14.2%
- Phosphorus: 7.7%
- UV Absorbance: 1.9 at wavelength 260–280 nm
Note: For research use only. Not intended for therapeutic or diagnostic applications.
It is also an excellent substrate for deoxyribonuclease or DNA polymerase. The solution contains sheared, single-stranded DNA molecules used to block the non-specific attachment of probe DNA to the surface of a membrane (Southern) or to increase yeast transformation efficiency. Solution vial should be stored at –20°C (–4°F). Expiration date is 6 months from the date of opening.
MDL Number:
MFCD00130922 CAS Number: 9007-49-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...