mLINE® Single Channel Mechanical Pipettors, Variable Volume, Sartorius
Supplier: Sartorius
725010 725020 725030 725050 725060 725070 725080 725090 725010 725090 725020 725030 725050 725060 725070 725080
14005-976EA 1360.57 CAD
14005-976 47745-544 47745-545 47745-546 47745-548 47745-550 47745-552 89079-110 CA14005-976 CA89079-110 CA47745-544 CA47745-545 CA47745-546 CA47745-548 CA47745-550 CA47745-552 CA10033-092 CA10033-094 CA10033-096 CA10033-098 CA10033-100
mLINE® Single Channel Mechanical Pipettors, Variable Volume, Sartorius
Pipettes Micropipettes
Sartorius mLINE® Mechanical Pipettors are an advanced and ergonomic choice for quality-conscious users who perform long and repetitive manual pipetting. Designed for repetitive and long-lasting procedures in order to prevent work-related hand, arm, and shoulder disorders or Repetitive Strain Injury (RSI).
  • Excellent ergonomics to protect from injuries with exceptionally light pipetting and tip ejection forces
  • High accuracy and precision, including in repetitive, long-lasting procedures
  • Minimized risk of contamination with Safe-Cone Filters and full
More Product Information