Finnpipette™ F1 Variable Volume Pipettes, Thermo Scientific
Supplier: Thermo Fisher Scientific
4641010N 4641020N 4641030N 4641040N 4641050N 4641060N 4641070N 4641080N 4641100N 4641110N 4641120N 4641130N 4641140N 4641010N 4641020N 4641030N 4641040N 4641050N 4641070N 4641110N 4641120N 4641090N 4641130N 4641140N
89080-413EA 911.7 CAD
89080-413 89080-415 89080-417 89080-419 89080-421 89080-423 89080-425 89080-427 89080-429 89080-431 89080-433 89126-243 89126-245 CA89080-413 CA89080-415 CA89080-417 CA89080-419 CA89080-421 CA89080-425 CA89080-431 CA89080-433 CA89080-569 CA89126-243 CA89126-245
Finnpipette™ F1 Variable Volume Pipettes, Thermo Scientific
Pipettes Micropipettes
The enhanced volume adjustment on Thermo Scientific™ Finnpipette™ F1 Variable Volume Pipettes is now silent and effortless with textured adjustment knob for a firmer feel when setting volumes.
  • Silent volume adjustment
  • Adjustable finger rest
  • Lightweight design
  • Secure volume lock
  • Increased air boost
  • Five-year warranty
The adjustable finger rest, and lightweight design maximizes comfort without sacrificing performance. Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI. Enjoy the new silentMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable