Ovation® QS, Digital Adjustable, Single Channel Pipettors, VistaLab
Supplier: VistaLab
1057-0020 1057-0200 1057-1000 1057-0002 1057-0010 1057-0100 1057-0999
14220-670EA 1374 CAD
14220-670 14220-674 14220-676 14220-678 14220-680 14220-672 93000-284
Ovation® QS, Digital Adjustable, Single Channel Pipettors, VistaLab
Pipettes Micropipettes
Revolutionary ergonomic design addresses the issues of posture, repetition, and force to significantly reduce strain and pain that occur from pipetting.
  • Adjustable hook provides custom comfort for left- or right-handed users
  • Tips are acquired with only a gentle push; audible click ensures a secure seal
  • Energy transfer button uses energy captured during tip acquisition to eject tip with minimal thumb force
  • Special filtered nozzle keeps liquid out of pipettor body
Five frequently-used aspirating and dispensing volumes can be easily programmed and recalled for use by pressing a single button. Other volume settings can be selected at precise increments throughout the pipettor’s volume range. Pipettor stands securely on counter, eliminating the need for racks and the risk of contamination from tip touching the bench.
A low-force, form-fitting plunger allows the user to manually control the speed of dispensing and aspirating with minimal effort. In-lab calibration can be easily performed without tools, to compensate for environmental conditions and/or different liquid types. Disposable battery lasts for six months of typical use with no recharging.
Available in six color-coded models for transferring liquid volumes from 0.2 to 1000 µl.
Design allows pipetting to be performed with a natural posture and minimal muscular movement: low hand elevation eases stress in the shoulder and neck, neutral wrist angle eliminates uncomfortable extension and radial deviation movements in the arm, and relaxed grip improves hand endurance.
Ordering information: The start-up kit includes the 2 to 20 µl, 20 to 200 µl, and 100 to 1000 µl volume pipettors. The kit also includes three racks of tips.