Eppendorf® Research® Plus Single-Channel Variable Volume Pipettors
Supplier: Eppendorf
3123000012 3123000020 3123000098 3123000039 3123000047 3123000055 3123000101 3123000063 3123000071 3123000080 3123000020 3123000012 3123000144
CA89125-292EA 941.95 CAD
CA89125-292 CA89125-294 CA89125-296 CA89125-298 CA89125-300 CA89125-302 CA89125-304 CA89125-306 CA89125-308 CA89125-310 89125-294 89125-292 CA77394-832
Eppendorf® Research® Plus Single-Channel Variable Volume Pipettors
Pipettes Micropipettes
These ergonomic pipettors are designed for simple, yet highly effective pipetting.
  • Color-coding on the operating button simplifies tip identification and selection
  • Secondary calibration option allows users to adjust to a wide variety of liquid handling requirements
  • Fully autoclavable devices can be easily disassembled for cleaning and decontamination
  • Ultra-light, corrosion-free piston runs softly and smoothly due to upgraded springs, guides, and seals
Pipettors feature a specially designed, spring-loaded tip cone to help reduce the force required for tip attachment and ejection. The rounded and elongated grip fits comfortably in the hand, enhancing stability and drastically reducing the strain resulting from repeated procedures. Operating and tip-ejector buttons have been enlarged and strategically positioned for simple access and operation. Volume adjustment requires only a few turns to reach desired settings, and features a display that reads to four digits.
They weigh up to 30% less than similar pipettors and feature optimal weight distribution to minimize stress to any one muscle area. The innovative mechanisms and specially developed precision components, such as springs, guides, and seals contribute to the overall ergonomic design of the device. The manual pipettors require up to 50% less force to operate when compared to similar pipettors. In addition, they are simple to service, autoclave, and calibrate in the laboratory.
Ordering information: Supplied with manufacturer’s three-year warranty. Each pipettor includes a factory Certificate of Conformity.