Kjeldahl Granules, Selenized, Hengar
Supplier: HENGAR INC
901200 901300
51137-046EA 680.46 CAD
51137-046 51137-068
Kjeldahl Granules, Selenized, Hengar
Digestion Systems Kjeldahl Tablets and Reagents
Granules greatly reduce time required for the Kjeldahl digestion, and ensure smooth boiling, both during digestion and subsequent distillation
Selenium is applied as a coating on the granules, as a convenient means to add to the digestion. Only two or three granules for each digestion flask is necessary.