Owl™ EasyCast™ Wide-Format Horizontal Electrophoresis Systems, Models D2 and D3, Thermo Scientific
Supplier: Thermo Fisher Scientific
D3-14 D2-BP
27373-200EA 1867.17 CAD
27373-200 27373-220 27373-327
Owl™ EasyCast™ Wide-Format Horizontal Electrophoresis Systems, Models D2 and D3, Thermo Scientific
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Screen multiple samples on a single agarose gel in less than half an hour
Systems incorporate the EasyCast™ design, wherein each side of the gel tray is fitted with a gasket, which allows the tray to fit snugly into the casting chamber. A leakproof seal is achieved without using tape. Built-in leveling ensures the casting of flat, uniformly thick gels.
Sample loading is enhanced using microwell format combs designed specifically for use with these systems. Samples may be loaded directly from a 96-well plate, 8 or 12 wells at a time.
The D3-14 system can run up to 200 samples (D2More Product Information