SOCOREX® ACURA® Manual 815 Single-Channel Pipettors With Tip Ejectors, Fixed Volume, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W810000 W810002 W810004 W810006 W810008 W810010 W810012 W810014 W810016 W810018 W810020 W810022 W810024 W810026 W810028 W810030 W810032 W810034 W810036 W810038 W810040 W810042 W810044 W810046 W810048 W810050 W810052 W810054 W810056 W810058 W810060 W810062
83009-092EA 829.9 CAD
83009-092 83009-094 83009-096 83009-098 83009-100 83009-102 83009-104 83009-106 83009-108 83009-110 83009-112 83009-114 83009-116 83009-118 83009-120 83009-122 83009-124 83009-126 83009-128 83009-130 83009-132 83009-134 83009-136 83009-138 83009-140 83009-142 83009-144 83009-146 83009-148 83009-150 83009-152 83009-154
SOCOREX® ACURA® Manual 815 Single-Channel Pipettors With Tip Ejectors, Fixed Volume, DWK Life Sciences
Pipettes Micropipettes
Lightweight pipettors are designed for precise, accurate pipetting of solutions, reagents, acids, serums, and other liquids, and for gel-sequencing applications.
  • Dispensing is accomplished with a single plunger stroke
  • Stainless steel and PVDF plunger assembly is chemically resistant
  • Autoclavable to 121 °C (250 °F) without disassembly or recalibration
A differentiated overshoot function and separate push button for tip ejection reduce dead volume and prevent accidental de-tipping. The glove-friendly JustTip™ adjustable tip ejector allows the use of most pipette tips.
Ordering information: Supplied with a manufacturer’s three-year warranty, including a two-year warranty on the original calibration. The 2000 µl pipettor includes an adapter for use with a Pasteur pipet. Pipettors 83009-148, -150, -152, and -154 are supplied with polypropylene fiber filters that may be used with polypropylene tips to prevent cross-contamination.