Acura® Pipettor, Fixed Volume, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
851343 851344
66187-651CS 1911.23 CAD
66187-651 66187-653
Acura® Pipettor, Fixed Volume, DWK Life Sciences
Pipettes Micropipettes
Ideal for bacteriology applications such as milk testing, this double volume instrument provides greater cost and time efficiency.
  • Unit features a fully integrated straw tip ejector
  • Sterile straw tips allow the user to pull samples from hard-to-reach areas
  • Tip length: 19 cm (7¹/₂")
  • Tips have a large opening to allow aspirating of small particles in liquid
  • Tips produce less waste than glass or plastic pipettes
Replaces glass and plastic pipettes and dilution bottles when performing sequential 1:10 dilutions. Dispenses 1 and 0.1 ml in one complete procedure.
Ordering information: Supplied with two-year warranty on original calibration and three-year manufacturer's warranty on parts and labor.