Eppendorf® Research® Plus Single-Channel Fixed Volume Pipettors
Supplier: Eppendorf
3124000024 3124000016 3124000040 3124000059 3124000067 3124000083 3124000091 3124000105 3124000113 3124000121 3124000032 3124000075
CA89125-312EA 562.3 CAD
CA89125-312 CA89125-314 CA89125-318 CA89125-320 CA89125-322 CA89125-326 CA89125-328 CA89125-330 CA89125-332 CA89125-334 CA89125-316 CA89125-324
Eppendorf® Research® Plus Single-Channel Fixed Volume Pipettors
Pipettes Micropipettes
These ergonomic pipettors are designed for simple, yet highly effective pipetting.
  • Color-coding on the operating button simplifies tip identification and selection
  • Volume adjustment requires only a few turns to reach desired settings
  • Features a display that reads to four digits
  • Secondary calibration option allows users to adjust to a wide variety of liquid handling requirements
  • Ultra-light, corrosion-free piston runs softly and smoothly due to upgraded springs, guides, and seals
  • Fully autoclavable devices can be easily disassembled for cleaning and decontamination
Pipettors feature a specially designed, spring-loaded tip cone to help reduce the force required for tip attachment and ejection. The rounded and elongated grip fits comfortably in the hand, enhancing stability and drastically reducing the strain resulting from repeated procedures. Operating and tip-ejector buttons have been enlarged and strategically positioned for simple access and operation.
They weigh up to 30% less than similar pipettors and feature optimal weight distribution to minimize stress to any one muscle area. The innovative mechanisms and specially developed precision components, such as springs, guides, and seals contribute to the overall ergonomic design of the pipettors.
The manual pipettors require up to 50% less force to operate when compared to similar devices. In addition, pipettors are simple to service, autoclave, and calibrate in the laboratory.
Ordering information: Supplied with manufacturer’s three-year warranty. Each pipettor includes a factory Certificate of Conformity.