Finnpipette™ F1 Fixed Volume Single-Channel Pipettes, Thermo Scientific
Supplier: Thermo Fisher Scientific
4651020N 4651030N 4651040N 4651050N 4651060N 4651070N 4651080N 4651090N 4651100N 4651110N 4651120N 4651130N 4651140N 4651020N 4651030N 4651050N 4651060N 4651070N 4651080N 4651090N 4651100N 4651110N 4651120N 4651130N 4651140N
89080-439EA 682.18 CAD
89080-439 89080-441 89080-443 89080-445 89080-447 89080-449 89080-451 89080-453 89080-455 89080-457 89080-459 89126-251 89126-253 CA89080-439 CA89080-441 CA89080-445 CA89080-447 CA89080-449 CA89080-451 CA89080-453 CA89080-455 CA89080-457 CA89080-459 CA89126-251 CA89126-253
Finnpipette™ F1 Fixed Volume Single-Channel Pipettes, Thermo Scientific
Pipettes Micropipettes
Great for set protocols and QA environments, Thermo Scientific™ Finnpipette™ F1 Fixed Volume Pipettes deliver a light, smooth pipetting action and an adjustable finger rest to maximize your comfort.
  • Smooth pipetting action
  • Adjustable finger rest
  • Lightweight design
  • Five-year warranty
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI. The 120° adjustable finger rest, for both right- and left-handed operator comfort, allows the hand to relax during pipetting cycles.
A 150% increase inMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable