VWR® Modular Heating Blocks for Microcentrifuge Tubes
Supplier: VWR International
12985-048 13259-000 13259-002 13259-286 12985-050 12985-044
12985-048EA 362.82 CAD
12985-048 13259-000 13259-002 13259-286 12985-050 12985-044
VWR® Modular Heating Blocks for Microcentrifuge Tubes
Block Heaters Blocks
Constructed from a solid anodized aluminum block.
  • Now with Best-in-Class, Exclusive 5-Year Warranty
The close contact of tubes to block walls allows for maximum heat retention. Single block size.