VWR® Modular Heating Sand Baths
Supplier: VWR International
13259-262 13259-264 13259-266 13259-268 13259-270
13259-262EA 730.54 CAD
13259-262 13259-264 13259-266 13259-268 13259-270
VWR® Modular Heating Sand Baths
Block Heaters Blocks
For use in heating blocks, these sand baths are ideal for irregular vessels.
  • Now with Best-in-Class, Exclusive 5-Year Warranty
Stainless steel construction for superior corrosion resistance. Designed to hold sand, shot, or nonvolatile liquids. Right angle thermometers (61220-740 and 61220-742) have an accuracy rating of ± 1°C.