UVP Transilluminators, Visi-White™, Analytik Jena
Supplier: Analytik Jena US
95-0214-01 95-0208-01 95-0214-02
21475-398EA 3247.34 CAD
21475-398 74690-036 74690-040
UVP Transilluminators, Visi-White™, Analytik Jena
Transilluminators White Light Transilluminators
View film negatives and positives, autoradiographs, X-ray film, microtiter plates, and stained gels
Glass surface is permanently double safety-sealed into the base for durability. Scratch- and chemical-resistant work surface can be easily cleaned. Housing is finished with an epoxy-powder paint for easy cleaning and maintenance.
Certifications: The 115 V models are ETL and ETL-C approved. The 230V models are CE marked.
Ordering information: Manufacturer’s two-year warranty on all models.