Automation Pipette Tips for QIAGEN® Workstations, Porex BioProducts
Supplier: Thermo Scientific
180-96R 181-96R BLK-180-96R BLK-180-96RS BLK-181-96R BLK-181-96RS
46300-468CS 1448.33 CAD
46300-468 46300-474 46300-478 46300-480 46300-482 46300-484
Automation Pipette Tips for QIAGEN® Workstations, Porex BioProducts
Pipette Tips
Compatible with QIAGEN® robotic workstations.
High quality, flexible polypropylene to ensure proper fit for accuracy and precision.
Available in two volume sizes, 300 and 1100 µl. Tips are available with filter and in two different colors, natural and black.