Automation Pipette Tips for Tecan® Workstations, Porex BioProducts
Supplier: Porex
173-96R 171-96R BLKF17596RS1000 BLK-175-96R BLK-175-96RS 175-96RS BLK-172-96RS BLK-173-96RS 172-96R 175-96R 173-96RS 171 LMF172-96RS-50 LMF171-96RS-10 172-96RS
46300-408CS 1445.76 CAD
46300-408 46300-398 46300-420 46300-388 46300-410 46300-440 46300-386 46300-428 46300-418 46300-404 46300-426 46300-424 46300-414 46300-422 46300-400
Automation Pipette Tips for Tecan® Workstations, Porex BioProducts
Pipette Tips
Tips are compatible with, but not limited to, the Tecan® Genesis workstation.
Volumes range from 10 to 1000 µl. Available in two different colors, natural and black. Also available with or without filter. The flexible polypropylene design provides optimal fit for the workstation. Sterile tips are tray-packaged.