Automation Pipette Tips for Zymark® Tempo™ Workstations, Porex BioProducts
Supplier: Thermo Scientific
160-96R 160-96RS 159-96R 159-96RS
46300-326CS 1047.98 CAD
46300-326 46300-328 46300-336 46300-338
Automation Pipette Tips for Zymark® Tempo™ Workstations, Porex BioProducts
Pipette Tips
Tips are compatible with the Zymark automated liquid handling systems.
Designed to ensure reliable operation with RapidPlate® 96, RapidPlate® 96/384, and all Micro Assay modules and workstations. Racked tips have a rigid box design for reliable tip attachment and individual compartments for each tip, preventing cross contamination.