Copper sulfate 1.053 Specific Gravity at 25°C (77°F) for Hemoglobin Estimation in Blood Donor Screening of Men and Women

Supplier: Ricca Chemical

R2330000-4C R2330000-500C R2330000-1C R2330000-20F
RC23301EA 273.34 CAD
RC23301 RC233016 RC233032 RC23305
Copper sulfate 1.053 Specific Gravity at 25°C (77°F) for Hemoglobin Estimation in Blood Donor Screening of Men and Women
Copper(II) sulphate solutions

For hemoglobin estimation in blood donor screening of men and women


Specific gravity is 1.053 at 25°C (77°F). Packaged in a glass amber container. Has a shelf life of 24 months.

Formula: CuSO₄·5H₂O
MDL Number: MFCD00149681
CAS Number: 7758-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR