Unimodule MGPI and METD Series Accessories, Envirco®
Supplier: Envirco
10229 10230 10231 10232 10336
VF-10229EA 867.75 CAD
VF-10229 VF-10230 VF-10231 VF-10232 VF-10336
Unimodule MGPI and METD Series Accessories, Envirco®
Fume Hoods and Enclosures Laminar Flow Cabinets
Gooseneck faucets are standard laboratory type with valve located above the work deck
Terminate below the work deck with a 0.95cm (3/8") IPS female outlet. Air, gas, or vacuum labcock is deck-mounted. For use with built-in work decks only. Duplex electrical outlets are surface-mounted, rated at 15A, and provided with separate 2.4m (8') three-conductor cord with molded plug. Stainless steel sink is suspended in work deck opening by 3.8cm (11/2") lip around top edge of unit and features a centrally located 8.9cm (31/2") stainless steel basket drain. Air velocity gauge is ball-type, mounted on control panel.