KIMBLE® KONTES® ULTRA-WARE® Funnel/Support Assemblies, 47 mm Glass and PTFE Face Supports, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
953755-0000 953780-0000
KT953755-0000EA 589 CAD
KT953755-0000 KT953780-0000
KIMBLE® KONTES® ULTRA-WARE® Funnel/Support Assemblies, 47 mm Glass and PTFE Face Supports, DWK Life Sciences
Filters Membrane Filter Units
This 47 mm vacuum filtration apparatus is designed to handle larger volumes of liquids for analysis of particulate or biological contamination.
Units have coarse porosity, fritted-glass filter support bases, available plain or with PTFE-faced funnel and support base to permit autoclaving with filter in place. An aluminum clamp sandwiches filter of choice between funnel and base. Connection to KT953760-0000 1 L filter flask (available as an accessory) is made with a silicone stopper.
Also for use with Millipore® filtering manifolds.
Order Now
SPECIFICATIONS
Environmentally Preferable