Socorex™ Dosys™ 164/174 Self-Refilling Premium Syringes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
851371 851372 851373 851375 W852014 W852016 W852018 W852020
80078-636CS 1136.83 CAD
80078-636 80078-638 80078-640 80078-642 89044-880 89044-882 89044-884 89044-886
Socorex™ Dosys™ 164/174 Self-Refilling Premium Syringes, DWK Life Sciences
Pipettes Repetitive Pipettors
Self-refilling automatic syringes with spring-loaded plunger allow repetitive delivery of aliquots from a single reservoir.
Corrosion-resistant stainless steel plungers offer a high degree of chemical resistance with no O-ring to replace. Handle design eliminates fatigue during extended operations. Two-ring or pistol-grip handles are available. Detachable valve system has corrosion-proof springs and stainless steel check-valve balls to ensure a repetitive accuracy of ±1%. Replaceable ground glass barrel with Viton® washers resists corrosion. Luer lock nozzle. Autoclavable.