Agarose

Supplier: Cytiva

17-0468-01 17-0468-01
CA95016-848LEA 693.45 CAD
95016-848 CA95016-848L
Agarose
Agarose

Agarose with low electroendosmosis (EEO) is recommended for most DNA/RNA applications.


Agarose IEF is suitable for isoelectric focusing of large molecules and is the gel of choice for many IEF applications and immunochemical detection methods.


If separated nucleic acids are to be used as substrates for enzymes (e.g., ligase and restriction endonucleases), a special highly purified agarose such as Agarose NA should be used.

MDL Number: MFCD00081294
CAS Number: 9012-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR