Accessories for UVP ChemStudio SA2 Imager with BioLite MultiSpectral Light Source Promotion, Analytik Jena
Supplier: Analytik Jena US
85-0006-01 85-0002-01 85-0007-02 85-0005-01
21475-522EA 253.72 CAD
21475-522 21475-523 21475-529 21475-521
Accessories for UVP ChemStudio SA2 Imager with BioLite MultiSpectral Light Source Promotion, Analytik Jena
Transilluminators UV Transilluminators
Gel-Cutter provides an edge for cutting and removal of gel material in one easy movement
Gel Ruler has centimeter marking fluoresce under UV wavelengths.
Clear plastic Gel-Ruler provides centimeter markings that fluoresce when exposed to 365 nm and 302 nm UV wavelengths for DNA analysis. Gel-Scooper, made of strong acrylic with a beveled edge, is designed to transfer gels easily from electrophoresis equipment to viewing equipment. Gel-Trays are made of UV-transmitting Plexiglas® acrylic and can be used for moving gels to the transilluminator. Trays protect transilluminator’s filterMore Product Information