Biotix Disposable Reagent Reservoirs, Biotix
Supplier: Biotix
SR-0100-5SNM SR-0050-1SC SR-0025-B3WM SR-0100-1SC SR-0025-5SWM SR-0055-1SNM SR-0025-13SWM SR-0100-5SC SR-0100-1SNM SR-0025-1SWM SR-0055-5SNM SR-0100-BC SR-0025-53SWM SR-0100-BNM SR-0025-BWM SR-0055-BNM SR-0050-5SC SR-0050-BC
89125-668CS 140.61 CAD
89125-668 89125-678 89125-676 89511-190 89125-674 89125-672 89125-670 89511-202 89511-188 89511-200 89511-206 89511-204 89511-194 89511-192 89511-186 89511-198 89511-184 89511-196
Biotix Disposable Reagent Reservoirs, Biotix
Reagent Reservoirs
Biotix disposable reagent reservoirs are designed for use in multi-channel pipetting applications and can accommodate up to 12-channel pipettors. Angled sides and a V-shaped bottom allow for maximum recovery of reagents. All reservoirs feature internal graduated volume lines and pour-spouts on the corners. Choose from opaque white polyvinyl chloride (PVC) or high clarity polystyrene (PS) in several packaging configurations.
Biotix disposable reagent reservoirs are designed for use in multi-channel pipetting applications and can accommodate up to 12-channel pipettors
Biotix Reagent reservoirs are certified free of DNA, DNase, RNase, and endotoxins (pyrogens). Manufactured under stringent controls in an ISO 9001 certified facility. Every lot is tested for engineering compliance and accuracy. All raw materials are traceable.
How Biotix Reservoirs can help in the lab:
Designed for use in multi-channel pipetting applications and fully compatible with 12-channel pipettors
Angled sides and a V-shaped bottom allow for maximum reagent recovery
Manufactured with internal volume graduation lines and pour-spouts on the corners
Available in polyvinyl chloride (PVC) or clear polystyrene (PS)
Angled sides and a V-shaped bottom allow for maximum recovery of reagents. All reservoirs feature internal graduated volume lines and pour-spouts on the corners. Choose from opaque white polyvinyl chloride (PVC) or high clarity polystyrene (PS) in several packaging configurations.
Certifications: CE certified.
Ordering information: Reagent reservoirs are available pre-sterile, nonsterile, bulk and individually-wrapped.