96-Well Microplates, Polypropylene, Greiner Bio-One
Supplier: Greiner Bio-One
650203 650204 650205 650206 650209 650261 651203 651205 651206 651209 651261 655204 655205 655206 655209 655261 650207 650201 651201 655201
82050-638CS 354.37 CAD
82050-638 82050-640 82050-642 82050-644 82050-646 82050-648 82050-668 82050-670 82050-672 82050-674 82050-676 82050-678 82050-778 82050-780 82050-782 82050-784 82050-786 89041-278 82050-636 82050-662 82050-774
96-Well Microplates, Polypropylene, Greiner Bio-One
Microplates HTS Microplates
These 96-well microplates are manufactured from ClearChoice™ virgin polypropylene resin and are produced with uniform external dimensions and tolerances to meet ANSI recommendations for compatibility with equipment and robotics.
  • Virgin polypropylene construction
  • Alphanumeric coding
  • Free of RNase, DNase, and human DNA
  • Nonpyrogenic
  • Stackable
  • Autoclavable
These plates are biologically inert, exhibit low binding characteristics, offer high temperature tolerance (–196 to 121 °C/–321 to 250 °F), and are resistant to many standard laboratory chemicals
Microplates feature alphanumerically coded wells. Round-bottom plates, conical-bottom plates, or flat-bottom plates enable complete sample recovery. Chimney well plates feature a chimney-like arrangement in which each well stands on its own to reduce the possibility of cross-contamination, and feature individual rims that rise above the surface of the plate to aid in sealing by cap mat, thermal sealer, or adhesive sealer. Stackable and autoclavable. Plates are supplied without lids. Dimensions: 128L×86W×15H mm.
Plates are free of detectable RNase, DNase, human DNA, and pyrogens. All plates have a footprint conforming to the ANSI/SBS 1-2004 Standard.
Black plates are ideal for fluorescence polarization.