Multi-Well Reagent Reservoirs
Supplier: Excel Scientific
RSVL175N10 RSV0175N10
89049-026CS 673.84 CAD
89049-026 89049-028 89049-030 89049-032
Multi-Well Reagent Reservoirs
Reagent Reservoirs
Reusable, large-capacity reservoirs feature a V-shaped basin that is compatible with both eight- and twelve-channel pipettors
  • Available with or without lids
  • Constructed of clear, chemical-resistant polypropylene for constant sample visibility
  • Molded graduation marks provide volume indication at 25, 50, 75, 100, 125, and 150mL
  • Autoclavable
The removable lids latch tightly for secure storage, and offer simple access that can be manipulated with one hand. Reservoirs are stackable with lids closed, and a perimeter ridge on each lid prevents sliding. A reinforced base eliminates wobbling and the threat of spilling.
They provide an environmentally sound alternative to disposable reservoirs, allow for fewer refills, and ensure minimal sample waste.