CELLSTAR® Tissue Culture Plates, Greiner Bio-One
Supplier: Greiner Bio-One
675083 675086 675090 675180 781098 650160 650161 650162 650185 651160 651161 651162 651180 655073 655079 655083 655087 655088 655090 655098 655160 655161 655162 655163 655185 781073 781079 781080 781086 781091 781092 781093 781165 781182 782073 782078 782080 782086 782092 782093 783092 783160 784080 784086 655182 782180 781090 781080-20 655090-99 781090-99 650180 655086 655180 651182
82050-048CS 295.1 CAD
82050-048 82050-050 82050-052 82050-064 82050-076 82050-625 82050-626 82050-628 82050-634 82050-655 82050-656 82050-658 82050-661 82050-722 82050-732 82050-736 82050-744 82050-746 82050-748 82050-758 82050-763 82050-764 82050-766 82050-768 82050-772 82051-266 82051-276 82051-278 82051-280 82051-282 82051-286 82051-288 82051-302 82051-304 82051-376 82051-390 82051-392 82051-394 82051-396 82051-398 82051-424 82051-438 82051-458 82051-460 82051-596 89047-272 89085-314 89096-044 89135-048 89135-050 82050-633 82050-742 82050-771 75799-866
CELLSTAR® Tissue Culture Plates, Greiner Bio-One
Microplates Cell Culture Plates
CELLSTAR® format sterile cell culture microplates are available tissue culture treated for the culture of anchorage-dependent cells, or untreated for suspension culture applications.
  • Designed to minimize wicking and reduce air bubble formation
  • Clear polystyrene film bottom
  • Chimney well design reduces cross-contamination
  • Certain models feature smaller working volumes to reduce sample usage
Multiwell culture plates come with a variety of well shapes
The 384-well plates with a maximum well volume of 30µL are special Small Volume™ plates that feature far smaller working volumes than typical 384-well plates, reducing sample usage while avoiding the instrument compatibility issues introduced by 1536-well plates. The wells on Small Volume™ plates feature conical well geometry; wells have a round top and a flat bottom, resembling a truncated cone.
Plates with µClear® bottoms feature a polystyrene microthin film bottom for clear imaging or viewing and minimized autofluorescence. HiBase plates are suitable for use with top-reading instruments, while LoBase plates are suitable for use with bottom-reading instruments. Dimensions: 128 L x 86 W mm (5 x 3⅜").
All plates are RNase- and DNase-free, noncytotoxic (EN ISO 10993), and nonpyrogenic. All plates have a Sterility Assurance Level (SAL) of 10-3 and have a footprint conforming to the ANSI/SBS 1-2004 Standard.