384-Well Small Volume™ Assay Plates, Greiner Bio-One
Supplier: Greiner Bio-One
784075 784076 784077 784101 788095 788096 788101 784075-25 784076-25
82051-442CS 467.75 CAD
82051-442 82051-444 82051-450 82051-456 82051-462 82051-484 82051-486 89085-324 89085-328 89085-330
384-Well Small Volume™ Assay Plates, Greiner Bio-One
Microplates HTS Microplates
Microplates provide high-throughput assay miniaturization in a 384-well system, thereby avoiding the instrument compatibility issues introduced by 1536-well plates while still reducing sample usage.
  • Suited for high-throughput assays
  • Free of RNase, DNase, and human DNA
  • Nonpyrogenic
  • Supplied sterile
  • Conical well geometry
All plates are manufactured from virgin polystyrene resin, except for the black UV-Star® 384-Well Microplates, which is constructed of cycloolefin copolymer (COC) for better chemical resistance and detections in lower UV wavelengths
Order Now
SPECIFICATIONS
Environmentally Preferable