384-Well No-Bottom Plates, Greiner Bio-One
Supplier: Greiner Bio-One
781000-06 788000-01
82051-262CS 587.55 CAD
82051-262 82051-544 89085-332
384-Well No-Bottom Plates, Greiner Bio-One
Microplates HTS Microplates
Standard and Small Volume™ LoBase plates are made from virgin polystyrene resin and molded without the bottom of the plate.
  • Smaller working volume reduces sample usage
  • Chimney-well design helps prevent cross-contamination
  • Virgin polystyrene construction
  • Molded without a bottom
The Small Volume format features far smaller working volumes than typical 384-well plates, reducing sample usage while avoiding the instrument compatibility issues introduced by 1536-well plates. The LoBase format features a chimney-like arrangement of wells in which each well stands on its own to reduce the possibility of cross-contamination. Chimney wells also feature individual rims that rise above the surface of the plate to aid in sealing by cap mat, thermal sealer, or adhesive sealer.