384-Well Clear Polystyrene Microplates, Greiner Bio-One
Supplier: Greiner Bio-One
781061 781101 781162 781185 781186 781061-25
82051-264CS 218.83 CAD
82051-264 82051-298 82051-300 82051-306 82051-308 89137-766
384-Well Clear Polystyrene Microplates, Greiner Bio-One
Microplates HTS Microplates
Microplates are manufactured from clear virgin polystyrene resin selected for high optical clarity and are designed to meet industry standard dimension specifications to facilitate compatibility with microplate handling and robotics.
  • Clear virgin polystyrene construction
  • High degree of optical clarity
  • Stackable
  • Designed to minimize air bubble formation
  • Flat bottoms and rounded tops
The MICROLON™ 600 format is available in plate 82051-264 for consistent performance in binding assays
Plate 82051-264 has a Sterility Assurance Level (SAL) of 10-3. Plates 82051-306 and -308 are supplied with lids. All plates are free of detectable RNase, DNase, human DNA, and pyrogens. They have a footprint conforming to the ANSI/SBS 1-2004 Standard.
Wells have flat bottoms and rounded square tops (square with round corners) designed to minimize wicking and reduce air bubble formation, while increasing volume capacity and surface area. Plates are stackable.