Pierce™ Reacti-Bind™ Streptavidin Coated 384-Well Plates, Thermo Scientific
Supplier: Thermo Scientific
15405 15407 15504 15505 15506
CAPI15405EA 1271.99 CAD
CAPI15405 CAPI15407 CAPI15504 CAPI15505 CAPI15506
Pierce™ Reacti-Bind™ Streptavidin Coated 384-Well Plates, Thermo Scientific
Microplates Immunology Microplates and Strips
Plates are ideal for binding small hydrophilic molecules that typically exhibit poor binding directly to polystyrene.
  • Low nonspecific binding
  • Easy and gentle immobilization of biotin-containing conjugates
  • No denaturing of the protein component of a conjugate upon binding
  • Pre-blocked with Blocker™ BSA or SuperBlock™ blocking buffer and ready-to-use
  • High binding capacity plates provides wider detection range and better curve linearity for small biotinylated ligands
Pierce™ streptavidin coated PS microplates are ideal for binding assays usingMore Product Information