Nunc® MicroWell™ 60- and 72-Well Mini Trays, Thermo Scientific
Supplier: Thermo Fisher Scientific
472400 438733 439225 470378 452256
CA62407-382CS 2333.42 CAD
CA62407-382 CA62409-296 CA62409-297 CA62407-126 CA62407-127
Nunc® MicroWell™ 60- and 72-Well Mini Trays, Thermo Scientific
Microplates Immunology Microplates and Strips
These Terasaki-style mini trays are ideal for serotyping, HLA applications, microlymphocytotoxicity, cell cloning studies, protein crystallization, and sequencing reactions.
Tray wells have a special polystyrene surface treatment to ensure enhanced mixing of reagents and samples in each well, even when frozen and thawed. Treatment ensures a limited affinity to antibodies even in high concentrations and limited protein adsorption. The optical clarity of the trays allows them to be viewed with an inverted microscope. Trays feature large raised letters for easy identification. Wells featureMore Product Information