ComboPlate™ 24-Well Protein Crystallization Plates, Greiner Bio-One
Supplier: Greiner bio-one
ComboPlate™
662150 662050
89047-256CS 578.6 CAD
89047-256 82050-890
ComboPlate™ 24-Well Protein Crystallization Plates, Greiner Bio-One
Microplates Crystallisation Plates
Made of virgin high-clarity resin, these universal microplates are designed for hanging and sitting drop vapor diffusion within a standard footprint.
  • Virgin resin construction
  • High degree of optical clarity
  • Compact footprint
  • Flat, raised rings minimize spills and cross-contamination
CrystalBridge™ well inserts fit into individual wells for sitting drop experiments and can be transferred to a different well mid-experiment
The distortion-free flat bottom provides excellent optical properties for microscopic inspection of crystal growth. Flat, raised rings around each well minimize spills and cross-contamination, and allow for sealing. Each plate comes with a slightly raised protective lid to prevent scratches and shifting of the seal during stacking or transportation. Plates are available in polystyrene (PS) for clarity or polyolefin for low birefringence (LBR). LBR plates are specially designed for use with polarized light. All plates have a footprint conforming to the ANSI/SBS 1-2004 Standard.
Maximum well volume with integrated insert: 45µL. Round coverslips made of siliconized glass, or VIEWseal™ clear adhesive tape can be used to seal each reservoir.