Streptavidin-Coated Plates, Greiner Bio-One
Supplier: Greiner Bio-One
781990 781995 781996 781997
82051-364CS 6361.33 CAD
82051-364 82051-366 82051-368 82051-370
Streptavidin-Coated Plates, Greiner Bio-One
Microplates Cell Culture Plates
Ideal for molecular biology applications, these plates feature low coating variance, high biotin binding capacity, and low streptavidin leaching.
  • Polystyrene construction
  • Available in a variety of clear or colored bottoms
  • Chimney-well design helps prevent cross-contamination
The very tight bond between streptavidin and biotin can be utilized to attach biotin-labeled proteins, nucleic acids, or other compounds to the microplate. Plates include a “Best Before” date and lot number on each pack. Available with black, white, clear, or µClear® bottom. The µClear® plates feature a reduced background, increasing sensitivity to 340nm. The 96-well plates feature a chimney-well design to prevent cross-contamination. 781990-589 and 781997-589 have blocker #5.