CELLCOAT® Cell Culture Microplates, Greiner Bio-One
Supplier: Greiner Bio-One
655930 655940 655944 655945 655946 655950 655956 657930 657940 657950 662930 662940 662950 781940 781944 781945 781946 781947 781950 655936 781936 781948 781956 655955 655948 781930 665940-005 662940-005 657950-005 665950-005 662950-005 657940-005
82050-798CS 543.31 CAD
82050-798 82050-800 82050-802 82050-804 82050-806 82050-810 82050-812 82050-848 82050-850 82050-852 82050-904 82050-906 82050-908 82051-352 82051-355 82051-356 82051-358 82051-360 82051-363 89047-252 89047-266 89047-268 89047-270 89131-870 89047-254 89218-660 75795-982 75795-992 75795-998 75796-000 75796-002 75796-008
CELLCOAT® Cell Culture Microplates, Greiner Bio-One
Microplates Cell Culture Plates
Convenient, ready-to-use polystyrene cell culture plates are coated with extracellular matrix (ECM) proteins or synthetic polypeptides.
  • Polystyrene construction
  • Compact footprint
  • Supplied with lid
  • Convenient and ready to use
These molecules provide an inert scaffolding that participates directly and indirectly in the regulation of cell growth and proliferation. Ideal for serum-free and serum-reduced cultivation or for differentiation of individual cell types. These multiwell tissue culture plates are consistently coated with Collagen Type 1 (from Rat Tail Tendon), Poly-D-Lysine or Poly-L-Lysine (both synthetic), and feature a “Best Before” date and lot number. All plates are supplied with lids and have a footprint conforming to the ANSI/SBS 1-2004 Standard.
Dimensions: 128L x 86W mm