Neptune® 96-Well PCR Plates, Biotix
Supplier: Biotix
63300617 63300626 63300634
89092-253CS 1163.78 CAD
89092-253 89126-692 89126-694
Neptune® 96-Well PCR Plates, Biotix
Microplates PCR Plates
PCR plates are molded from high performance polypropylene and feature thin walls for efficient heat transfer
Plate seals work with all Neptune™ plates for seamless integration between research and storage. PCR plate seal (89092-218) is optically clear with an aggressive seal that is perfect for reducing evaporation experienced during thermal cycling. Aluminum sealing tape (89092-220) is chemically resistant, pierceable, and ideal for PCR and sample storage. Plate sealing tape (89092-222) is non-pierceable polypropylene film. This film is not recommended for sealing raised rim plates.
Compatible with many of the latest thermocyclers on the market, these PCR plates are available in 96-well low profile, non-skirted, semi-skirted, and fully skirted versions. Refer to PCR plate compatibility chart for fit with specific thermocyclers.
Well thickness: 0.381 mm (1/64")
Certifications: CE certified. IVD labeled. Certified free of DNA, DNase, RNase, and endotoxins (pyrogens). Manufactured under stringent controls in an ISO 9001 certified facility. Every lot is tested for precision assuring maximum quality and compatibility. All raw materials are traceable.