Neptune® Specialty-Use Pipette Tips
Supplier: Biotix
63302171 63300555 63302174
89140-154CS 1260.23 CAD
89140-154 89140-156 89140-158
Neptune® Specialty-Use Pipette Tips
Pipette Tips
These precision universal pipette tips are made from virgin polypropylene.
  • Autoclavable
  • Good sustainability
  • Superior stability, firm placement
  • Maximum space saving design
Eco-friendly empty pipette tips trays are manufactured from recycled polypropylene. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%.
The extended length facilitates access to deep vessels. The thin wall design ensures an optimal seal and easy ejection with industry leading pipettors. Eco-friendly racks are made from recycled polypropylene and utilize a unique compact packaging configuration that reduces the carbon footprint.
Certifications: Certified free of RNase, DNase, pyrogens, and PCR Inhibitors. Manufactured under stringent controls in an ISO 9001 certified facility. Each and every lot is tested for accuracy and precision, ensuring superior quality and compatibility. All raw materials are controlled.