Neptune® S³ Low Retention Pipette Tips
Supplier: Biotix
63302157 63300508 63302158 63302167 63300544 63302163 63302164 63302165 63300534 63302160 63300520 63302161 63302178 63300567 63302177
89140-096CS 1039.03 CAD
89140-096 89140-098 89140-100 89140-106 89140-128 89140-130 89140-884 89140-888 89140-892 89140-898 89168-732 89168-734 89168-736 89168-758 89168-760 89425-648
Neptune® S³ Low Retention Pipette Tips
Pipette Tips
The exclusive S³ polymer delivers to within 0.1% of indicated volume, eliminating sample hold-up and ensuring a more precise application.
  • Autoclavable
  • Good sustainability
  • Superior stability, firm placement
  • Maximum space saving design
Eco-friendly empty pipette tips trays are manufactured from recycled polypropylene. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%.
This proprietary technology is essential for quantitative assays where sample retention can hinder accurate results. Universal pipette tips areMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Sustainable Packaging,Safer Human and Environmental Health